missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8015 Arizona Calibration Standard Mixture 240 10000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30320702
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320702 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320702 Supplier Dr. Ehrenstorfer Supplier No. DREA50000240DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material No. 2 Diesel Oil∣SAE 10W-30 motor oil
CAS 68334-30-5, 329050-13-7
Analyte or Component Names No. 2 Diesel Oil∣SAE 10W-30 motor oil
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) No. 2 Diesel Oil 10000 μg/mL∣SAE 10W-30 motor oil 10000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.