missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 634 Carbamate Mixture 395 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Butylate∣Cycloate∣EPTC∣Molinate∣Pebulate∣Vernolate |
| CAS | 2008-41-5, 1134-23-2, 759-94-4, 2212-67-1, 1114-71-2, 1929-77-7 |
| Analyte or Component Names | Butylate∣Cycloate∣EPTC∣Molinate∣Pebulate∣Vernolate |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Butylate 1000 μg/mL∣Cycloate 1000 μg/mL∣EPTC 1000 μg/mL∣Molinate 1000 μg/mL∣Pebulate 1000 μg/mL∣Vernolate 1000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction