missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 625/8270 Composites Mixture 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2-Methylnaphthalene∣2-Nitroaniline∣3-Nitroaniline∣4-Chloroaniline∣4-Nitroaniline∣Aniline∣Benzyl alcohol∣Carbazole∣Dibenzofuran∣Pyridine |
| CAS | 91-57-6, 88-74-4, 99-09-2, 106-47-8, 100-01-6, 62-53-3 |
| Analyte or Component Names | 2-Methylnaphthalene∣2-Nitroaniline∣3-Nitroaniline∣4-Chloroaniline∣4-Nitroaniline∣Aniline∣Benzyl alcohol∣Carbazole∣Dibenzofuran∣Pyridine |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2-Methylnaphthalene 2000 μg/mL∣2-Nitroaniline 2000 μg/mL∣3-Nitroaniline 2000 μg/mL∣4-Chloroaniline 2000 μg/mL∣4-Nitroaniline 2000 μg/mL∣Aniline 2000 μg/mL∣Benzyl alcohol 2000 μg/mL∣Carbazole 2000 μg/mL∣Dibenzofuran 2000 μg/mL∣Pyridine 2000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction