missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 624/625 Tuning Standards Mixture 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30309120
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309120 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309120 Supplier Dr. Ehrenstorfer Supplier No. DREA50000281DI
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Benzidine∣4,4'-DDT∣Decafluorotriphenylphosphine∣Pentachlorophenol
CAS 92-87-5, 50-29-3, 5074-71-5, 87-86-5
Analyte or Component Names 4,4'-Benzidine∣4,4'-DDT∣Decafluorotriphenylphosphine∣Pentachlorophenol
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4,4'-Benzidine 1000 μg/mL∣4,4'-DDT 1000 μg/mL∣Decafluorotriphenylphosphine 1000 μg/mL∣Pentachlorophenol 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.