missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 610 Additions PAH Mixture 445 5-100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 3-Methylcholanthrene∣7H-Dibenzo[c,g]carbazole∣Acenaphthene∣Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(j)fluoranthene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenz(A,J)Acridine∣Dibenzo(a,e)pyrene∣Dibenzo(a,h)anthracene∣Dibenzo(A,I)Pyrene∣Dibenzo[a,h]acridine∣Dibenzo[a,h]pyrene∣Fluoranthene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Naphthalene∣Phenanthrene∣Pyrene |
| CAS | 56-49-5, 194-59-2, 83-32-9, 208-96-8, 120-12-7, 56-55-3 |
| Analyte or Component Names | 3-Methylcholanthrene∣7H-Dibenzo[c,g]carbazole∣Acenaphthene∣Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(j)fluoranthene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenz(A,J)Acridine∣Dibenzo(a,e)pyrene∣Dibenzo(a,h)anthracene∣Dibenzo(A,I)Pyrene∣Dibenzo[a,h]acridine∣Dibenzo[a,h]pyrene∣Fluoranthene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Naphthalene∣Phenanthrene∣Pyrene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Benzo(k)fluoranthene 5 μg/mL∣3-Methylcholanthrene 10 μg/mL∣7H-Dibenzo[c,g]carbazole 10 μg/mL∣Benz[a]anthracene 10 μg/mL∣Benzo(j)fluoranthene 10 μg/mL∣Benzo[a]pyrene 10 μg/mL∣Benzo[b]fluoranthene 10 μg/mL∣Benzo[ghi]perylene 10 μg/mL∣Chrysene 10 μg/mL∣Dibenz(A,J)Acridine 10 μg/mL∣Dibenzo(a,e)pyrene 10 μg/mL∣Dibenzo(a,h)anthracene 10 μg/mL∣Dibenzo(A,I)Pyrene 10 μg/mL∣Dibenzo[a,h]acridine 10 μg/mL∣Dibenzo[a,h]pyrene 10 μg/mL∣Fluoranthene 10 μg/mL∣Indeno[1,2,3-cd]pyrene 10 μg/mL∣Pyrene 10 μg/mL∣Acenaphthene 100 μg/mL∣Acenaphthylene 100 μg/mL∣Anthracene 100 μg/mL∣Fluorene 100 μg/mL∣Naphthalene 100 μg/mL∣Phenanthrene 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?