missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 555 Chlorinated Acids Mixture 464 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4,5-Trichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣4-Nitrophenol∣Dinoseb∣MCPA∣Mecoprop∣Pentachlorophenol |
| CAS | 93-76-5, 51-36-5, 94-82-6, 100-02-7, 88-85-7, 94-74-6 |
| Analyte or Component Names | 2,4,5-Trichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣4-Nitrophenol∣Dinoseb∣MCPA∣Mecoprop∣Pentachlorophenol |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4,5-Trichlorophenoxyacetic acid 1000 μg/mL∣3,5-Dichlorobenzoic acid 1000 μg/mL∣4-(2,4-Dichlorophenoxy)butyric acid 1000 μg/mL∣4-Nitrophenol 1000 μg/mL∣Dinoseb 1000 μg/mL∣MCPA 1000 μg/mL∣Mecoprop 1000 μg/mL∣Pentachlorophenol 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?