missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 552 Halogenated Acetic Mixture 80-800 μg/mL in Methyl tert-butyl ether, Dr. Ehrenstorfer

Product Code. 30324480
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30324480 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30324480

Brand: Dr. Ehrenstorfer DREGA09000362MB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromoacetic acid∣Bromochloroacetic acid∣Bromodichloroacetic acid∣Chloroacetic acid∣Dibromoacetic acid∣Dibromochloroacetic acid∣Dichloroacetic acid∣Tribromoacetic acid∣Trichloroacetic acid (TCA)
CAS 79-08-3, 5589-96-8, 71133-14-7, 79-11-8, 631-64-1, 5278-95-5
Analyte or Component Names Bromoacetic acid∣Bromochloroacetic acid∣Bromodichloroacetic acid∣Chloroacetic acid∣Dibromoacetic acid∣Dibromochloroacetic acid∣Dichloroacetic acid∣Tribromoacetic acid∣Trichloroacetic acid (TCA)
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix MTBE
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Dichloroacetic acid 80 μg/mL∣Bromoacetic acid 120 μg/mL∣Bromochloroacetic acid 120 μg/mL∣Bromodichloroacetic acid 120 μg/mL∣Dibromoacetic acid 120 μg/mL∣Dibromochloroacetic acid 120 μg/mL∣Trichloroacetic acid (TCA) 200 μg/mL∣Chloroacetic acid 800 μg/mL∣Tribromoacetic acid 800 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.