missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 537 Internal Standard Primary Dilution Standard, 1-4 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2-(N-Methylperfluorooctanesulfonamido)acetic acid D3 (N-methyl D3),Perfluorooctanesulfonic Acid 13C4 (1,2,3,4-13C4) Sodium,Perfluorooctanoic Acid-13C2 |
| Packaging | Bottle |
| CAS | 1400690-70-1, 960315-53-1, 864071-08-9 |
| Concentration or Composition Notes | 2-(N-Methylperfluorooctanesulfonamido)acetic acid D3 (N-methyl D3) 4 μg/mL,Perfluorooctanesulfonic Acid 13C4 (1,2,3,4-13C4) Sodium 3 μg/mL,Perfluorooctanoic Acid-13C2 1 μg/mL |
| Analyte or Component Names | 2-(N-Methylperfluorooctanesulfonamido)acetic acid D3 (N-methyl D3),Perfluorooctanesulfonic Acid 13C4 (1,2,3,4-13C4) Sodium,Perfluorooctanoic Acid-13C2 |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solution Type | Mixture |
| Type | Multi-component Standard |
| Solvent or Matrix | Methanol/Water |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?