missing translation for 'onlineSavingsMsg'
Learn More

Dr. Ehrenstorfer EPA Method 537.1 Surrogate Primary Dilution Standard 1 to 4 μg/mL in Methanol:Water

Product Code. 31784136
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31784136 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31784136 Supplier Dr. Ehrenstorfer Supplier No. DREA50000791MW
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Certified reference materials available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality.

  • Certified reference materials
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluoro-2-propoxypropanoic Acid 13C3 (Propionic Acid 13C3), 2-(N-Ethylperfluorooctanesulfonamido)acetic Acid D5 (N-Ethyl D5), Perfluorohexanoic Acid 13C2 (1,2-13C2), Perfluorodecanoic Acid 13C2 (1,2-13C2)
CAS 3030247-97-0, 1265205-97-7, 960315-47-3, 960315-50-8
Shipping Condition 20°C
Concentration 1 to 4 μg/mL
Certifications/Compliance UKAS ISO 17034
Solution Type Mixture
Solvent or Matrix Methanol/Water
Industry Type Environmental Testing
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.