missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 537.1 PFAS Mixture 100 μg/mL in Methanol:Water, Dr. Ehrenstorfer

Product Code. 30312733
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312733 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312733 Supplier Dr. Ehrenstorfer Supplier No. DREA50000152MW
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid potassium∣2-(N-Ethylperfluorooctanesulfonamido)acetic acid∣2-(N-Methylperfluorooctanesulfonamido)acetic acid∣3H-Perfluoro-4,8-dioxanonanoic acid∣9-Chloro-perfluoro-3-oxanonanesulfonic acid potassium∣Perfluoro-2-propoxypropanoic acid (PFPrOPrA)∣Perfluorobutanesulfonic acid∣Perfluorodecanoic acid∣Perfluorododecanoic acid∣Perfluoroheptanoic acid∣Perfluorohexanesulfonic acid∣Perfluorohexanoic acid∣Perfluorononanoic acid∣Perfluorooctanesulfonic acid∣Perfluorooctanoic acid∣Perfluorotetradecanoic acid∣Perfluorotridecanoic acid∣Perfluoroundecanoic acid
CAS 83329-89-9, 2991-50-6, 2355-31-9, 919005-14-4, 73606-19-6, 13252-13-6
Analyte or Component Names 11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid potassium∣2-(N-Ethylperfluorooctanesulfonamido)acetic acid∣2-(N-Methylperfluorooctanesulfonamido)acetic acid∣3H-Perfluoro-4,8-dioxanonanoic acid∣9-Chloro-perfluoro-3-oxanonanesulfonic acid potassium∣Perfluoro-2-propoxypropanoic acid (PFPrOPrA)∣Perfluorobutanesulfonic acid∣Perfluorodecanoic acid∣Perfluorododecanoic acid∣Perfluoroheptanoic acid∣Perfluorohexanesulfonic acid∣Perfluorohexanoic acid∣Perfluorononanoic acid∣Perfluorooctanesulfonic acid∣Perfluorooctanoic acid∣Perfluorotetradecanoic acid∣Perfluorotridecanoic acid∣Perfluoroundecanoic acid
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol/Water
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid potassium 100 μg/mL∣2-(N-Ethylperfluorooctanesulfonamido)acetic acid 100 μg/mL∣2-(N-Methylperfluorooctanesulfonamido)acetic acid 100 μg/mL∣3H-Perfluoro-4,8-dioxanonanoic acid 100 μg/mL∣9-Chloro-perfluoro-3-oxanonanesulfonic acid potassium 100 μg/mL∣Perfluoro-2-propoxypropanoic acid (PFPrOPrA) 100 μg/mL∣Perfluorobutanesulfonic acid 100 μg/mL∣Perfluorodecanoic acid 100 μg/mL∣Perfluorododecanoic acid 100 μg/mL∣Perfluoroheptanoic acid 100 μg/mL∣Perfluorohexanesulfonic acid 100 μg/mL∣Perfluorohexanoic acid 100 μg/mL∣Perfluorononanoic acid 100 μg/mL∣Perfluorooctanesulfonic acid 100 μg/mL∣Perfluorooctanoic acid 100 μg/mL∣Perfluorotetradecanoic acid 100 μg/mL∣Perfluorotridecanoic acid 100 μg/mL∣Perfluoroundecanoic acid 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.