missing translation for 'onlineSavingsMsg'
Learn More

Dr. Ehrenstorfer EPA Method 533 Isotope Dilution Standard 0.5 to 2 μg/mL in Methanol:Water

Product Code. 31784133
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31784133 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31784133 Supplier Dr. Ehrenstorfer Supplier No. DREA50000790MW
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified reference materials available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality.

  • Certified reference materials
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluoro-2-propoxypropanoic Acid 13C3 (Propionic Acid 13C3), Perfluoroundecanoic Acid 13C7 (1,2,3,4,5,6,7-13C7), 1H,1H,2H,2H-Perfluorohexanesulfonic Acid 13C2 (1,2-13C2) Sodium, Perfluorohexanoic Acid 13C5 (1,2,3,4,6-13C5), Perfluorooctanoic acid 13C8, Perfluorobutanoic acid 13C4, Perfluoroheptanoic Acid 13C4 (1,2,3,4-13C4), Perfluorononanoic acid 13C9, Perfluorooctanesulfonic acid 13C8 sodium, Perfluoropentanoic acid 13C5, Perfluorodecanoic acid 13C6 (1,2,3,4,5,6-13C6), Perfluorobutanesulfonic Acid 13C3 (2,3,4-13C3) Sodium, Perfluorohexanesulfonic Acid 13C3 (1,2,3-13C3) Sodium, 1H,1H,2H,2H-Perfluorooctanesulfonic Acid 13C2 (1,2-13C2) Sodium, 1H,1H,2H,2H-Perfluorodecanesulfonic Acid 13C2 (1,2-13C2) Sodium, Perfluorododecanoic Acid (1,2-13C2)
CAS 3030247-97-0, 2708218-88-4, 2328024-54-8, 1350614-84-4, 1017281-29-6, 2328024-55-9
Shipping Condition 20°C
Concentration 0.5 to 2 μg/mL
Certifications/Compliance UKAS ISO 17034
Solution Type Mixture
Solvent or Matrix Methanol/Water
Industry Type Environmental Testing
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.