missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 531.1 Carbamate Pesticide Mixture 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30310764
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30310764

Brand: Dr. Ehrenstorfer DREGA09000948ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Methiocarb∣Methomyl∣Oxamyl∣Propoxur
CAS 116-06-3, 1646-88-4, 1646-87-3, 63-25-2, 1563-66-2, 16655-82-6
Analyte or Component Names Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Methiocarb∣Methomyl∣Oxamyl∣Propoxur
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Aldicarb 100 μg/mL∣Aldicarb-sulfone 100 μg/mL∣Aldicarb-sulfoxide 100 μg/mL∣Carbaryl 100 μg/mL∣Carbofuran 100 μg/mL∣Carbofuran-3-hydroxy 100 μg/mL∣Methiocarb 100 μg/mL∣Methomyl 100 μg/mL∣Oxamyl 100 μg/mL∣Propoxur 100 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.