missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.3 UCMR 4 Surrogate Mixture 500 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30309738
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309738 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309738 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000264AC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dimethyl-2-nitrobenzene (2-Nitro-m-xylene)∣Benzo(a)pyrene D12∣Triphenylphosphate
CAS 81-20-9, 63466-71-7, 115-86-6
Analyte or Component Names 1,3-Dimethyl-2-nitrobenzene (2-Nitro-m-xylene)∣Benzo(a)pyrene D12∣Triphenylphosphate
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,3-Dimethyl-2-nitrobenzene (2-Nitro-m-xylene) 500 μg/mL∣Benzo(a)pyrene D12 500 μg/mL∣Triphenylphosphate 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.