missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.3 UCMR 4 Pesticide Mixture 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30326745
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326745 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326745 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000262ME
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-HCH∣Chlorpyrifos∣Dimethipin∣Ethoprophos∣Oxyfluorfen∣Permethrin∣Profenofos∣Tebuconazole∣Tribufos
CAS 319-84-6, 2921-88-2, 55290-64-7, 13194-48-4, 42874-03-3, 52645-53-1
Analyte or Component Names alpha-HCH∣Chlorpyrifos∣Dimethipin∣Ethoprophos∣Oxyfluorfen∣Permethrin∣Profenofos∣Tebuconazole∣Tribufos
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) alpha-HCH 1000 μg/mL∣Chlorpyrifos 1000 μg/mL∣Dimethipin 1000 μg/mL∣Ethoprophos 1000 μg/mL∣Oxyfluorfen 1000 μg/mL∣Permethrin 1000 μg/mL∣Profenofos 1000 μg/mL∣Tebuconazole 1000 μg/mL∣Tribufos 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.