missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.3 PAH Mixture 500 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30321663
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321663 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321663 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000345AC
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acenaphthylene∣Adipic Acid Bis(2-ethylhexyl) Ester∣Anthracene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Dibutyl phthalate∣Diethyl phthalate∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Phenanthrene∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene
CAS 208-96-8, 103-23-1, 120-12-7, 56-55-3, 207-08-9, 50-32-8
Analyte or Component Names Acenaphthylene∣Adipic Acid Bis(2-ethylhexyl) Ester∣Anthracene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Dibutyl phthalate∣Diethyl phthalate∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Phenanthrene∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Acenaphthylene 500 μg/mL∣Adipic Acid Bis(2-ethylhexyl) Ester 500 μg/mL∣Anthracene 500 μg/mL∣Benz[a]anthracene 500 μg/mL∣Benzo(k)fluoranthene 500 μg/mL∣Benzo[a]pyrene 500 μg/mL∣Benzo[b]fluoranthene 500 μg/mL∣Benzo[ghi]perylene 500 μg/mL∣Chrysene 500 μg/mL∣Dibenzo(a,h)anthracene 500 μg/mL∣Dibutyl phthalate 500 μg/mL∣Diethyl phthalate 500 μg/mL∣Fluorene 500 μg/mL∣Indeno[1,2,3-cd]pyrene 500 μg/mL∣Isophorone 500 μg/mL∣Phenanthrene 500 μg/mL∣Phthalic acid, benzylbutyl ester 500 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 500 μg/mL∣Phthalic acid, bis-methyl ester 500 μg/mL∣Pyrene 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.