missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.2 PCB Congeners Mixture 500 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30328636
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328636 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328636 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000340AC
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PCB 47 (2,2',4,4'-Tetrachlorobiphenyl)∣PCB 5 (2,3-Dichlorobiphenyl)∣PCB No. 1∣PCB No. 154∣PCB No. 171∣PCB No. 201∣PCB No. 29∣PCB No. 88
CAS 2437-79-8, 16605-91-7, 2051-60-7, 60145-22-4, 52663-71-5, 40186-71-8
Analyte or Component Names PCB 47 (2,2',4,4'-Tetrachlorobiphenyl)∣PCB 5 (2,3-Dichlorobiphenyl)∣PCB No. 1∣PCB No. 154∣PCB No. 171∣PCB No. 201∣PCB No. 29∣PCB No. 88
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) PCB 47 (2,2',4,4'-Tetrachlorobiphenyl) 500 μg/mL∣PCB 5 (2,3-Dichlorobiphenyl) 500 μg/mL∣PCB No. 1 500 μg/mL∣PCB No. 154 500 μg/mL∣PCB No. 171 500 μg/mL∣PCB No. 201 500 μg/mL∣PCB No. 29 500 μg/mL∣PCB No. 88 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.