missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.2 PAH Mixture 386 500 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30319832
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319832 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319832 Supplier Dr. Ehrenstorfer Supplier No. DREA50000386AC
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dimethyl-2-nitrobenzene (2-Nitro-m-xylene)∣Acenaphthene D10∣Chrysene D12∣Perylene D12∣Phenanthrene D10∣Pyrene-d10∣Triphenylphosphate
CAS 81-20-9, 15067-26-2, 1719-03-5, 1520-96-3, 1517-22-2, 1718-52-1
Analyte or Component Names 1,3-Dimethyl-2-nitrobenzene (2-Nitro-m-xylene)∣Acenaphthene D10∣Chrysene D12∣Perylene D12∣Phenanthrene D10∣Pyrene-d10∣Triphenylphosphate
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,3-Dimethyl-2-nitrobenzene (2-Nitro-m-xylene) 500 μg/mL∣Acenaphthene D10 500 μg/mL∣Chrysene D12 500 μg/mL∣Perylene D12 500 μg/mL∣Phenanthrene D10 500 μg/mL∣Pyrene-d10 500 μg/mL∣Triphenylphosphate 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.