missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 524.2 Revision Mixture 2000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30313007
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313007 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313007 Supplier Dr. Ehrenstorfer Supplier No. DREA50000043ME
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dichloropropanone-2∣1-Chlorobutane∣2-Butanone∣2-Hexanone∣2-Nitropropane∣4-Methyl-2-pentanone (MIBK)∣Acetone∣Acrylic acid methyl ester∣Acrylonitrile∣Allylchloride∣Carbon disulfide∣Chloroacetonitrile∣Diethylether∣Hexachloroethane∣Methacrylic acid-ethyl ester∣Methacrylic acid-methyl ester∣Methacrylonitrile∣Methyl iodide∣Methyl tert-butyl ether∣Nitrobenzene∣Propionitrile∣Tetrahydrofuran∣trans-1,4-Dichloro-2-butene
CAS 513-88-2, 109-69-3, 78-93-3, 591-78-6, 79-46-9, 108-10-1
Analyte or Component Names 1,1-Dichloropropanone-2∣1-Chlorobutane∣2-Butanone∣2-Hexanone∣2-Nitropropane∣4-Methyl-2-pentanone (MIBK)∣Acetone∣Acrylic acid methyl ester∣Acrylonitrile∣Allylchloride∣Carbon disulfide∣Chloroacetonitrile∣Diethylether∣Hexachloroethane∣Methacrylic acid-ethyl ester∣Methacrylic acid-methyl ester∣Methacrylonitrile∣Methyl iodide∣Methyl tert-butyl ether∣Nitrobenzene∣Propionitrile∣Tetrahydrofuran∣trans-1,4-Dichloro-2-butene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,1-Dichloropropanone-2 2000 mg/L∣1-Chlorobutane 2000 mg/L∣2-Butanone 2000 mg/L∣2-Hexanone 2000 mg/L∣2-Nitropropane 2000 mg/L∣4-Methyl-2-pentanone (MIBK) 2000 mg/L∣Acetone 2000 mg/L∣Acrylic acid methyl ester 2000 mg/L∣Acrylonitrile 2000 mg/L∣Allylchloride 2000 mg/L∣Carbon disulfide 2000 mg/L∣Chloroacetonitrile 2000 mg/L∣Diethylether 2000 mg/L∣Hexachloroethane 2000 mg/L∣Methacrylic acid-ethyl ester 2000 mg/L∣Methacrylic acid-methyl ester 2000 mg/L∣Methacrylonitrile 2000 mg/L∣Methyl iodide 2000 mg/L∣Methyl tert-butyl ether 2000 mg/L∣Nitrobenzene 2000 mg/L∣Propionitrile 2000 mg/L∣Tetrahydrofuran 2000 mg/L∣trans-1,4-Dichloro-2-butene 2000 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.