missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 515.2 Underivatized Mixture 404 100-500 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30314380
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314380 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314380 Supplier Dr. Ehrenstorfer Supplier No. DREA50000404ME
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,5-Trichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣Chlorthal-diacid∣Dichlorprop∣Dinoseb∣Pentachlorophenol
CAS 93-76-5, 51-36-5, 2136-79-0, 120-36-5, 88-85-7, 87-86-5
Analyte or Component Names 2,4,5-Trichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣Chlorthal-diacid∣Dichlorprop∣Dinoseb∣Pentachlorophenol
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4,5-Trichlorophenoxyacetic acid 100 μg/mL∣Chlorthal-diacid 100 μg/mL∣Dichlorprop 100 μg/mL∣Pentachlorophenol 100 μg/mL∣Dinoseb 200 μg/mL∣3,5-Dichlorobenzoic acid 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.