missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 5035, High Level Amber Vial, P/T MeOH, Tared, 5 mL, 72/cs, Dr. Ehrenstorfer

Product Code. 30328732
Change view
Click to view available options
Analyte or Component Names:
Methanol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Analyte or Component Names Unit Size
30328732 Methanol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328732 Supplier Dr. Ehrenstorfer Supplier No. DREO000013
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanol
CAS 67-56-1
Analyte or Component Names Methanol
Shipping Condition Room Temperature
Molecular Formula C H4 O
Certifications/Compliance ISO 17025
Synonym Methanol, Methyl alcohol
InChI Formula InChI=1S/CH4O/c1-2/h2H,1H3
SMILES CO
IUPAC Name methanol
Industry Type Environmental Testing
Molecular Weight (g/mol) 32.04
Formula Weight 32.0262 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.