missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 200.8: 10000 mg/L Ca, Na; 1000 mg/L K, Mg, VHG™

Product Code. 17928799
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17928799 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17928799 Supplier VHG Labs Supplier No. VHG1600683

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

EPA Method 200.8: 10000 mg/L Ca, Na; 1000 mg/L K, Mg
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium/Potassium/Calcium/Sodium
CAS 7439-95-4, 9-7-7440, 7440-70-2, 7440-23-5
Methods ICP/ICP-MS
Analyte or Component Names Magnesium/Potassium/Calcium/Sodium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Magnesium 1000 mg/L/Potassium 1000 mg/L/Calcium 10000 mg/L/Sodium 10000 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.