missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 200.8: 10 mg/L Al, As, Be, Cd, Co, Cr, Mn, Mo, Ni, Pb, Sb, Se, Tl, Th, U, V , VHG™

Product Code. 17909179
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909179 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17909179 Supplier VHG Labs Supplier No. VHG1600681
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

EPA Method 200.8: 10 mg/L Al, As, Be, Cd, Co, Cr, Mn, Mo, Ni, Pb, Sb, Se, Tl, Th, U, V
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Antimony/Arsenic/Beryllium/Cadmium/Chromium/Cobalt/Lead/Manganese/Molybdenum/Nickel/Selenium/Thallium/Thorium/Uranium/Vanadium
CAS 7429-90-5, 7440-36-0, 7440-38-2, 7440-41-7, 7440-43-9, 7440-47-3
Methods ICP/ICP-MS
Analyte or Component Names Aluminium/Antimony/Arsenic/Beryllium/Cadmium/Chromium/Cobalt/Lead/Manganese/Molybdenum/Nickel/Selenium/Thallium/Thorium/Uranium/Vanadium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Aluminium 10 mg/L/Antimony 10 mg/L/Arsenic 10 mg/L/Beryllium 10 mg/L/Cadmium 10 mg/L/Chromium 10 mg/L/Cobalt 10 mg/L/Lead 10 mg/L/Manganese 10 mg/L/Molybdenum 10 mg/L/Nickel 10 mg/L/Selenium 10 mg/L/Thallium 10 mg/L/Thorium 10 mg/L/Uranium 10 mg/L/Vanadium 10 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.