missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 1633 PFAS Mixture 10 μg/mL in Methanol:Water, Dr. Ehrenstorfer

Product Code. 30328582
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328582 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328582 Supplier Dr. Ehrenstorfer Supplier No. DREA50000753MW
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1,2,2H-Perfluorodecanesulfonic acid∣1,1,2,2H-Perfluorohexanesulfonic acid∣1,1,2,2H-Perfluorooctanesulfonic acid∣11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid potassium∣1H,1H,2H,2H-Perfluorodecanoic acid∣2-(N-Ethylperfluorooctanesulfonamido)acetic acid∣2-(N-Methylperfluorooctanesulfonamido)acetic acid∣2H,2H,3H,3H-Perfluorohexanoic acid∣2H,2H,3H,3H-Perfluorooctanoic Acid∣3H-Perfluoro-4,8-dioxanonanoic acid∣9-Chloro-perfluoro-3-oxanonanesulfonic acid potassium∣N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide∣N-Ethyl-N-(2-hydroxyethyl)perfluorooctylsulfonamide∣N-Methylperfluorooctanesulfonamide∣Perfluoro(2-ethoxyethane) sulfonic acid∣Perfluoro-2-propoxypropanoic acid (PFPrOPrA)∣Perfluoro-3,6-dioxaheptanoic acid (PFDHA)∣Perfluoro-3-methoxypropanoic acid (PFMOPrA)∣Perfluoro-4-methoxybutanoic acid (PFMOBA)∣Perfluorobutanesulfonic acid∣Perfluorobutanoic acid∣Perfluorodecane sulfonic acid∣Perfluorodecanoic acid∣Perfluorododecanesulfonic Acid Sodium∣Perfluorododecanoic acid∣Perfluoroheptanesulfonic acid∣Perfluoroheptanoic acid∣Perfluorohexanesulfonic acid∣Perfluorohexanoic acid∣Perfluorononanesulfonic acid sodium salt∣Perfluorononanoic acid∣Perfluorooctane sulfonamide PFOSA∣Perfluorooctanesulfonic acid∣Perfluorooctanoic acid∣Perfluoropentanesulfonic Acid∣Perfluoropentanoic acid∣Perfluorotetradecanoic acid∣Perfluorotridecanoic acid∣Perfluoroundecanoic acid∣Sulfluramid
CAS 39108-34-4, 757124-72-4, 27619-97-2, 83329-89-9, 812-70-4, 2991-50-6
Analyte or Component Names 1,1,2,2H-Perfluorodecanesulfonic acid∣1,1,2,2H-Perfluorohexanesulfonic acid∣1,1,2,2H-Perfluorooctanesulfonic acid∣11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid potassium∣1H,1H,2H,2H-Perfluorodecanoic acid∣2-(N-Ethylperfluorooctanesulfonamido)acetic acid∣2-(N-Methylperfluorooctanesulfonamido)acetic acid∣2H,2H,3H,3H-Perfluorohexanoic acid∣2H,2H,3H,3H-Perfluorooctanoic Acid∣3H-Perfluoro-4,8-dioxanonanoic acid∣9-Chloro-perfluoro-3-oxanonanesulfonic acid potassium∣N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide∣N-Ethyl-N-(2-hydroxyethyl)perfluorooctylsulfonamide∣N-Methylperfluorooctanesulfonamide∣Perfluoro(2-ethoxyethane) sulfonic acid∣Perfluoro-2-propoxypropanoic acid (PFPrOPrA)∣Perfluoro-3,6-dioxaheptanoic acid (PFDHA)∣Perfluoro-3-methoxypropanoic acid (PFMOPrA)∣Perfluoro-4-methoxybutanoic acid (PFMOBA)∣Perfluorobutanesulfonic acid∣Perfluorobutanoic acid∣Perfluorodecane sulfonic acid∣Perfluorodecanoic acid∣Perfluorododecanesulfonic Acid Sodium∣Perfluorododecanoic acid∣Perfluoroheptanesulfonic acid∣Perfluoroheptanoic acid∣Perfluorohexanesulfonic acid∣Perfluorohexanoic acid∣Perfluorononanesulfonic acid sodium salt∣Perfluorononanoic acid∣Perfluorooctane sulfonamide PFOSA∣Perfluorooctanesulfonic acid∣Perfluorooctanoic acid∣Perfluoropentanesulfonic Acid∣Perfluoropentanoic acid∣Perfluorotetradecanoic acid∣Perfluorotridecanoic acid∣Perfluoroundecanoic acid∣Sulfluramid
Shipping Condition -18°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol/Water
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,1,2,2H-Perfluorodecanesulfonic acid 10 μg/mL∣1,1,2,2H-Perfluorohexanesulfonic acid 10 μg/mL∣1,1,2,2H-Perfluorooctanesulfonic acid 10 μg/mL∣11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid potassium 10 μg/mL∣1H,1H,2H,2H-Perfluorodecanoic acid 10 μg/mL∣2-(N-Ethylperfluorooctanesulfonamido)acetic acid 10 μg/mL∣2-(N-Methylperfluorooctanesulfonamido)acetic acid 10 μg/mL∣2H,2H,3H,3H-Perfluorohexanoic acid 10 μg/mL∣2H,2H,3H,3H-Perfluorooctanoic Acid 10 μg/mL∣3H-Perfluoro-4,8-dioxanonanoic acid 10 μg/mL∣9-Chloro-perfluoro-3-oxanonanesulfonic acid potassium 10 μg/mL∣N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide 10 μg/mL∣N-Ethyl-N-(2-hydroxyethyl)perfluorooctylsulfonamide 10 μg/mL∣N-Methylperfluorooctanesulfonamide 10 μg/mL∣Perfluoro(2-ethoxyethane) sulfonic acid 10 μg/mL∣Perfluoro-2-propoxypropanoic acid (PFPrOPrA) 10 μg/mL∣Perfluoro-3,6-dioxaheptanoic acid (PFDHA) 10 μg/mL∣Perfluoro-3-methoxypropanoic acid (PFMOPrA) 10 μg/mL∣Perfluoro-4-methoxybutanoic acid (PFMOBA) 10 μg/mL∣Perfluorobutanesulfonic acid 10 μg/mL∣Perfluorobutanoic acid 10 μg/mL∣Perfluorodecane sulfonic acid 10 μg/mL∣Perfluorodecanoic acid 10 μg/mL∣Perfluorododecanesulfonic Acid Sodium 10 μg/mL∣Perfluorododecanoic acid 10 μg/mL∣Perfluoroheptanesulfonic acid 10 μg/mL∣Perfluoroheptanoic acid 10 μg/mL∣Perfluorohexanesulfonic acid 10 μg/mL∣Perfluorohexanoic acid 10 μg/mL∣Perfluorononanesulfonic acid sodium salt 10 μg/mL∣Perfluorononanoic acid 10 μg/mL∣Perfluorooctane sulfonamide PFOSA 10 μg/mL∣Perfluorooctanesulfonic acid 10 μg/mL∣Perfluorooctanoic acid 10 μg/mL∣Perfluoropentanesulfonic Acid 10 μg/mL∣Perfluoropentanoic acid 10 μg/mL∣Perfluorotetradecanoic acid 10 μg/mL∣Perfluorotridecanoic acid 10 μg/mL∣Perfluoroundecanoic acid 10 μg/mL∣Sulfluramid 10 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.