missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 1631, Stannous Chloride, 8 x 40g, Dr. Ehrenstorfer

Product Code. 30321525
Change view
Click to view available options
Quantity:
8 x 40 g
Unit Size:
320g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321525 8 x 40 g 320g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321525 Supplier Dr. Ehrenstorfer Supplier No. DREO000002
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stannous Chloride
CAS 7772-99-8
Analyte or Component Names Stannous Chloride
Shipping Condition Room Temperature
Molecular Formula SnCl2
Certifications/Compliance ISO 17025
Synonym Tin Dichloride, Dichlorotin, Tin Protochlorie, Tin Chloride, Tin (II) Chloride, Stannous Dichloride
InChI Formula InChI=1S/2ClH.Sn/h2*1H;/q;;+2/p-2
SMILES Cl[Sn]Cl
IUPAC Name dichlorotin
Industry Type Environmental Testing
Molecular Weight (g/mol) 189.61
Quantity 8 x 40 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.