missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 1625 Hydrocarbons Mixture 4000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | n-Decane∣n-Docosane∣n-Dodecane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Octadecane∣n-Tetradecane∣Octacosane∣Tetracosane∣Triacontane |
| CAS | 124-18-5, 629-97-0, 112-40-3, 112-95-8, 630-01-3, 544-76-3 |
| Analyte or Component Names | n-Decane∣n-Docosane∣n-Dodecane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Octadecane∣n-Tetradecane∣Octacosane∣Tetracosane∣Triacontane |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | n-Decane 4000 μg/mL∣n-Docosane 4000 μg/mL∣n-Dodecane 4000 μg/mL∣n-Eicosane 4000 μg/mL∣n-Hexacosane 4000 μg/mL∣n-Hexadecane 4000 μg/mL∣n-Octadecane 4000 μg/mL∣n-Tetradecane 4000 μg/mL∣Octacosane 4000 μg/mL∣Tetracosane 4000 μg/mL∣Triacontane 4000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?