missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Eotaxin Recombinant Rabbit Monoclonal Antibody (0Z5K4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557970
This item is not returnable.
View return policy
Description
Immunogen sequence: GSIPTSCCFI MTSKKIPNTL LKSYKRITNN RCTLKAIVFK TRLGKEICAD PKKKWVQDAT KHLDQKLQTP KP.
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications
| Eotaxin | |
| Recombinant Monoclonal | |
| 3.04 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P48298 | |
| CCL11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 26-97 of mouse CCL11/Eotaxin (NP_035460.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0Z5K4 | |
| Unconjugated | |
| CCL11 | |
| C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20292 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction