missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Eotaxin Recombinant Rabbit Monoclonal Antibody (0Z5K4)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557969

 View more versions of this product

Product Code. 30283108

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GSIPTSCCFI MTSKKIPNTL LKSYKRITNN RCTLKAIVFK TRLGKEICAD PKKKWVQDAT KHLDQKLQTP KP.

This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Eotaxin
Recombinant Monoclonal
3.04 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P48298
CCL11
Recombinant fusion protein containing a sequence corresponding to amino acids 26-97 of mouse CCL11/Eotaxin (NP_035460.1).
20 μL
Primary
Mouse
Antibody
IgG
ELISA, Western Blot
0Z5K4
Unconjugated
CCL11
C-C motif chemokine 11; C-C motif chemokine ligand 11; Ccl11; chemokine (C-C motif) ligand 11; eosinophil chemotactic protein; Eotaxin; Eotaxin 1; Eotaxin1; eotaxin-1; MGC22554; Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; small-inducible cytokine A11
Rabbit
Protein A
RUO
20292
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.