Learn More
PeproTech Eotaxin 3 Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Brand: PeproTech 500-P156G-100UG
Description
AA Sequence of recombinant protein: TRGSDISKTC CFQYSHKPLP WTWVRSYEFT SNSCSQRAVI FTTKRGKKVC THPRKKWVQK YISLLKTPKQ L. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human Eotaxin-3 (CCL26). Anti-Human Eotaxin-3 (CCL26)-specific antibody was purified by affinity chromatography employing an immobilized Human Eotaxin-3 (CCL26) matrix. Sandwich ELISA: To detect Human Eotaxin-3 (CCL26) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Eotaxin-3 (CCL26) (500-P156GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Eotaxin-3 (CCL26). Western Blot: To detect Human Eotaxin-3 (CCL26) by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Eotaxin-3 (CCL26) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Eotaxin-3 is a small cytokine belonging to the CC chemokine family that is also called CCL26, Chemokine (C-C motif) ligand 26. Chemokines play pivotal roles in health and disease by controlling migration and tissue localization of specific types of cells expressing their cognate receptors. Eotaxin-3 is chemotactic for eosinophils and basophils and elicits its effects by binding to the cell surface chemokine receptor CCR3. It is expressed by several tissues including heart, lung and ovary, and in endothelial cells that have been stimulated with the cytokine interleukin 4. This gene for chemokine is located on human chromosome 7. Eotaxin-3 induced migration of eosinophils and basophils only at a 10-fold higher concentration than eotaxin.
Specifications
| Eotaxin 3 | |
| Polyclonal | |
| Unconjugated | |
| CCL26 | |
| CC chemokine IMAC; C-C motif chemokine 26; C-C motif chemokine ligand 26; Ccl26; Ccl26l; chemokine (C-C motif) ligand 26; chemokine N1; Eotaxin3; eotaxin-3; IMAC; Macrophage inflammatory protein 4-alpha; MGC126714; MIP-4a; MIP-4alpha; MIP-4-alpha; PRO242; SCYA26; SCYA26MIP-4-alpha; small inducible cytokine A26; small inducible cytokine subfamily A (Cys-Cys), member 26; small-inducible cytokine A26; Thymic stroma chemokine-1; TSC-1; UNQ216; UNQ216/PRO242 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 10344 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9Y258 | |
| CCL26 | |
| E.coli-derived Recombinant Human Eotaxin-3 (CCL26). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.