Learn More
PeproTech Eotaxin 2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
Brand: PeproTech 500-P175GBT-50UG
Description
AA Sequence of recombinant protein: VTIPSSCCTS FISKKIPENR VVSYQLANGS ICPKAGVIFI TKKGHKICTD PKLLWVQRHI QKLDAKKNQP SKGAKAVRTK FAVQRRRGNS TEV. Preparation: Produced from sera of goats immunized with highly pure Recombinant Murine Eotaxin-2 (CCL24). Anti-Murine Eotaxin-2 (CCL24)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mEotaxin-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine Eotaxin-2 (500-P175G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mEotaxin-2. Western Blot: To detect mEotaxin-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mEotaxin-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P175GBT-1MG will be provided as 2 x 500 μg
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
Specifications
| Eotaxin 2 | |
| Polyclonal | |
| Biotin | |
| CCL24 | |
| CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 56221 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9JKC0 | |
| CCL24 | |
| E.coli-derived Recombinant Murine Eotaxin-2 (CCL24). | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.