missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin Y, pure, Indicator grade

Product Code. 10206310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10206310 100 g Glass bottle 100g
10368480 25 g Glass bottle 25g
10349183 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10206310 Supplier Thermo Scientific Chemicals Supplier No. 201931000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym 2';,4';,5';,7';-Tetrabromofluorescein, disodium salt, Eosin Yellowish, 2';, 4';, 5';
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Melting Point >300.0°C
Beilstein 19, IV, 2917
Merck Index 15, 3658
Solubility Information Solubility in water: freely soluble. Other solubilities: sparingly soluble in alcohol, insoluble in diethyl ether
Infrared Spectrum Authentic
Lambda Max 511 to 519nm
Formula Weight 691.85
Loss on Drying 10% max.
Grade Pure
Packaging Glass bottle
Specific Extinction 1100 min. (lambda max, 0.001M Na2CO3) on dried substance
Specific Gravity 1.018
UV Spectrometry Complies
Density 1.0180g/mL
Color Brown/Red
Quantity 100 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Eosin Y
Show More Show Less
compliance-icons
Product Identifier
  • Eosin Y
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acid red 87

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.