missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin Y, pure, certified

Product Code. 10577742 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10577742 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10577742 Supplier Thermo Scientific Chemicals Supplier No. 409430250
  • ONLINE EXCLUSIVE PRICE
    £29.70
    £26.65 / 25g
    Save £3.05
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17372-87-1 | C20H12Br4Na2O8 | 691.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

CAS 17372-87-1
Absorption Ratio 1.21 to 1.77
Dye Content 80% min.
Infrared Spectrum Authentic
Lambda Max 515 to 518nm
Packaging Glass bottle
Molecular Formula C20H12Br4Na2O8
MDL Number MFCD00005040
Beilstein 19, IV, 2917
Density 1.0180g/mL
Merck Index 15, 3658
Synonym 2',4',5',7'-Tetrabromofluorescein, disodium salt, Eosin Yellowish, C.I. 45380, 2', 4', 5'
Color Brown to Brown/Green
Quantity 25 g
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Physical Form Powder
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
Molecular Weight (g/mol) 691.85
PubChem CID 91886399
Formula Weight 691.85
Grade Pure
Chemical Name or Material Eosin Y
Show More Show Less
compliance-icons
Product Identifier
  • Eosin Y
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.