missing translation for 'onlineSavingsMsg'
Learn More

Eosin B, pure, spirit soluble, Thermo Scientific™

Product Code. 10154310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10154310 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10154310 Supplier Thermo Scientific Acros Supplier No. 199650250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56360-46-4
Molecular Formula C20H6Br2N2O9-2
MDL Number MFCD01104119
Beilstein 19, 233
Synonym C.I. 45400
InChI Key ZBQZBWKNGDEDOA-UHFFFAOYSA-L
Infrared Spectrum Authentic
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)[N+](=O)[O-])[O-])Br)Br)[O-])[N+](=O)[O-]
IUPAC Name 4',5'-dibromo-2',7'-dinitro-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
Molecular Weight (g/mol) 578.081
PubChem CID 7058116
ChEBI CHEBI:68617
Formula Weight 580.09
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Orange
Quantity 25 g
Physical Form Powder
Chemical Name or Material Eosin B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.