Learn More
Invitrogen™ EOMES Monoclonal Antibody (Dan11mag), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 14-4875-80
Description
Description: This Dan11mag antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Applications Reported: This Dan11mag antibody has been reported for use in immunohistochemistry of formalin-fixed paraffin embedded tissue sections, intracellular staining followed by flow cytometric analysis, and western blotting. (Fluorochrome conjugated Dan11mag is recommended for use in intracellular flow cytometry.). Applications Tested: This Dan11mag antibody has been tested by intracellular staining followed by flow cytometric analysis of mouse splenocytes and/or western blotting at 5 μg/mL. This Dan11mag antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded mouse tissue using low pH antigen retrieval at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
Specifications
| EOMES | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| O54839 | |
| Eomes | |
| Affinity chromatography | |
| RUO | |
| 13813 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Dan11mag | |
| Unconjugated | |
| Eomes | |
| C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
| Rat | |
| 25 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.