missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EOMES Monoclonal Antibody (21Mags8), eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Invitrogen™ 14-4876-80

 View more versions of this product

Product Code. 15227397

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Description: This 21Mags8 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Note: For detection of mouse Eomes, we recommend 12-4875 and 51-4875 and for detection of human Eomes, we recommend 51-4877 due to the brighter staining intensity of these products compared to clone 21Mags8. Applications Reported: This 21Mags8 antibody has been reported for use in flow cytometric analysis, and immunoblotting (WB). Applications Tested: This 21Mags8 antibody has been tested by western blot analysis of mouse splenocytes. As a starting concentration, this antibody can be used at 2 μg/mL. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EOMES
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O54839, O95936
Eomes
Affinity chromatography
RUO
13813, 8320
4°C
Liquid
Flow Cytometry, Western Blot
21Mags8
Unconjugated
Eomes
C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2
Rat
25 μg
Primary
Human, Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.