missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EOMES Antibody (CL15425) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
£321.00 - £459.00
Specifications
| Antigen | EOMES |
|---|---|
| Clone | CL15425 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279757
|
Novus Biologicals
NBP3-43667-100ul |
100 μL |
£459.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279603
|
Novus Biologicals
NBP3-43667-25ul |
25 μL |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EOMES Monoclonal antibody specifically detects EOMES in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)Specifications
| EOMES | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| PBS, pH 7.2, 40% glycerol | |
| 8320 | |
| IgG1 | |
| Protein A purified |
| CL15425 | |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
| This antibody was generated using a recombinant protein sequence of O95936, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title