missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EOGT Recombinant Rabbit Monoclonal Antibody (23GB3585), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553414
This item is not returnable.
View return policy
Description
The exact function of C3orf64 remains unknown.
Specifications
| EOGT | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5NDL0, Q5NDL2, Q8BYW9 | |
| Eogt | |
| A synthetic peptide of human AER61 (1-150AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3585 | |
| Unconjugated | |
| Eogt | |
| A130022J15Rik; Aer61; AER61 glycosyltransferase; AI447490; AOS4; AW214473; AW259391; C3orf64; EGF domain specific O-linked N-acetylglucosamine transferase; EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase; EGF domain-specific O-linked N-acetylglucosamine transferase; EGF-O-GlcNAc transferase; EOGT; EOGT1; Extracellular O-linked N-acetylglucosamine transferase; glycosyltransferase Aer61 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 101351, 285203, 494219 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction