missing translation for 'onlineSavingsMsg'
Learn More

Enzymatic Digest Casein Hydrolysate

Product Code. p-7076796 Shop All Thermo Scientific Products
Click to view available options
Quantity:
11.3 kg (25 lb.)
2.2 kg (5 lb.)
Unit Size:
2268g
25lb
This item is not returnable. View return policy

Product Code. 15895378

Brand: Thermo Scientific J12855.Q1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enzymatic digest of casein.

  • Synonym: Peptones
  • pH: 6.6 to 7.5
  • CAS: 65072-00-6
  • MDLNumber: MFCD00282824
TRUSTED_SUSTAINABILITY

Specifications

Name Casein hydrolysate
pH Range 6.6 to 7.5
Quantity 11.3 kg (25 lb.)
Packaging Plastic bottle
Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.