missing translation for 'onlineSavingsMsg'
Learn More

Lanopepden, MedChemExpress

Product Code. 16407927
Encompass_Preferred
Click to view available options
Quantity:
1 mg
2 mg
5 mg
Unit Size:
1mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16407927

Brand: Medchem Express HY12480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lanopepden (GSK 1322322) is a peptide deformylase inhibitor active against Staphylococcus aureus strains with MICs of 1 and 1 mg/L for ATCC 29213 and ATCC 25923 strain, respectively.

Lanopepden (GSK 1322322) is a peptide deformylase inhibitor active against Staphylococcus aureus strains with MICs of 1 and 1 mg/L for ATCC 29213 and ATCC 25923 strain, respectively.

Specifications

Chemical Name or Material Lanopepden
Physical Form Solid
CAS 1152107-25-9
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H34FN7O4
For Use With (Application) COVID-19-immunoregulation
Synonym GSK 1322322
SMILES FC1=C(N2CCN(CCOC3)[C@@]3([H])C2)N=C(C)N=C1NNC([C@H](CC4CCCC4)CN(C=O)O)=O
Molecular Weight (g/mol) 479.55
Formula Weight 479.55
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.