missing translation for 'onlineSavingsMsg'
Learn More

Enzalutamide Carboxylic Acid, TRC

Product Code. 30005297
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005297 2.5 mg 2.5mg
30006171 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005297 Supplier Toronto Research Chemicals Supplier No. TRCE5598002.5MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1242137-15-0
Molecular Formula C20H13F4N3O3S
Molecular Weight (g/mol) 451.39
Synonym 4-[3-[4-Cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-4-oxo-2-thioxo-1-imidazolidinyl]-2-fluoro-benzoic Acid
SMILES O=C(O)C1=CC=C(N(C(N2C3=CC=C(C#N)C(C(F)(F)F)=C3)=S)C(C)(C)C2=O)C=C1F

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C20H13F4N3O3S/c1-19(2)17(30)26(11-4-3-10(9-25)14(7-11)20(22,23)24)18(31)27(19)12-5-6-13(16(28)29)15(21)8-12/h3-8H,1-2H3,(H,28,29)
Chemical Name or Material Enzalutamide Carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.