Learn More
Environmental Cal Standard A (Primary): Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, U, V, Zn @ 10; Ca, Fe, K, Mg, Na @ 1000 μg/mL in 5% HNO3, tr. HF, tr. Tartaric Acid, VHG™
Product Code. 30003015
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003015 500 mL 500mL
17998039 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003015 Supplier VHG Labs Supplier No. VHGLCAL1A500
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Environmental Cal Standard A (Primary): Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, U, V, Zn @ 10; Ca, Fe, K, Mg, Na @ 1000 μg/mL in 5% HNO3, tr. HF, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL