missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences pENTRY-IBA51 Vector
GREENER_CHOICE

Product Code. 15603748 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15603748

Brand: IBA Lifesciences 54091001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

IBA Lifesciences GmbH Entry Vector pENTRY-IBA51 is required to generate Donor Vectors for subsequent Destination Vector generation.

  • StarGate Entry Vector is designed to insert the PCR product of a certain gene of interest (GOI) to generate a Donor Vector.

  • StarGate cloning via the pENTRY-IBA vector provides a tool for the systematic and rapid screen of the best working expression conditions for a certain GOI, where nothing is known from literature.

  • After initial pENTRY cloning, the GOI in the resulting Donor Vector can be easily transferred by a simple one-tube reaction into a multitude of expression vectors (Acceptor Vectors) providing different features.

Specifications

Marker Gene Kanamycin
Product Type Cloning Vector
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Protein Tag Untagged
Concentration 250 ng/μL
Quantity 5 μg
Vector pENTRY-IBA51
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado