missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port Right (Individual Location)

Product Code. 17199744 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17199744 HPP2200eco Each
17109654 HPP1060eco Each
17109794 HPP260eco Each
17119884 HPP750eco Each
17120195 IPP260eco Each
17120235 IPP410eco Each
17140275 IPP750eco Each
17159724 HPP1400eco Each
17159834 HPP410eco Each
17160135 IPP1400eco Each
17169694 HPP110eco Each
17170155 IPP2200eco Each
17180065 IPP1060eco Each
17190105 IPP110eco Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 17199744 Supplier Memmert™ Supplier No. HPP2200ecoF5
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Entry port with flap closure and custom TBC position in right side

  • Not in combination with illumination units
  • Moisture tight
  • Can be closed by flap and silicone stopper

Specifications

Color Clear
For Use With (Equipment) HPP2200eco
Diameter (Metric) 23 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.