missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port Right Centre/Top

Product Code. 17199874 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17199874 HPP750eco Each
17100195 IPP260eco Each
17100235 IPP410eco Each
17120275 IPP750eco Each
17139834 HPP410eco Each
17149694 HPP110eco Each
17160065 IPP1060eco Each
17170105 IPP110eco Each
17189644 HPP1060eco Each
17189784 HPP260eco Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17199874 Supplier Memmert™ Supplier No. HPP750ecoF3
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For introducing connections at the side and can be closed by flap

  • Standard position right centre/top
  • Not for modelsize 110 with light module

Specifications

Color Clear
For Use With (Equipment) HPP750eco
Diameter (Metric) 23 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.