missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port Rear (Individual Position)

Product Code. 17119654 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17119654 HPP1060eco Each
17119794 HPP260eco Each
17129884 HPP750eco Each
17130195 IPP260eco Each
17130235 IPP410eco Each
17150275 IPP750eco Each
17169834 HPP410eco Each
17179694 HPP110eco Each
17190065 IPP1060eco Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 17119654 Supplier Memmert™ Supplier No. HPP1060ecoF6
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Entry port with flap closure and custom position TBC in the back

  • Not in combination with illumination units
  • Moisture tight
  • Can be closed by flap and silicone stopper

Specifications

Color Clear
For Use With (Equipment) HPP1060eco
Diameter (Metric) 23 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.