Learn More
Memmert™ Entry Port Left Centre/Centre
Product Code. 17109834 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17109834 HPP410eco Each
17119694 HPP110eco Each
17130065 IPP1060eco Each
17140105 IPP110eco Each
17159644 HPP1060eco Each
17159784 HPP260eco Each
17169874 HPP750eco Each
17170225 IPP410eco Each
17180185 IPP260eco Each
17190265 IPP750eco Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17109834 Supplier Memmert™ Supplier No. HPP410ecoF0
Only null left
Add to basket
Add to basket
For introducing connections at the side. Moisture tight and can be closed by flap and silicone stopper
  • Standard position left centre/centre
  • Not for model size 260 with light module
  • Not for model size 110 with light module
Specifications
Description Moisture tight
For Use With (Equipment) HPP410eco