Learn More
Memmert™ Entry Port 57 mm for IN75
Product Code. 17598005 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN75 Incubator
Unit Size:
Each
Product Code. 17598005
Brand: Memmert™ IN75F8
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 57 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IN75 Incubator
Type Entry Port
Diameter (Metric) 57 mm