missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Entry Port 23 mm, Standard Position Left Side Centre/Top, For Un110Pa

Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.

Brand:  Memmert™ UN110paF1

Additional Details : Weight : 0.01100kg

Product Code. 16324267

  • £182.00 / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

  • Entry port with clear diameter
  • For Memmert Ovens
Specifications

Specifications

Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Stainless Steel
23 mm
Entry Port
0.9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers