Learn More
Memmert™ Entry Port 23 mm for IN75, Custom Position TBC Left Side
Product Code. 17558005 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN75 Incubator
Unit Size:
Each
Product Code. 17558005
Brand: Memmert™ IN75F4
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in left side
Specifications
For Use With (Equipment) IN75 Incubator
Type Entry Port
Diameter (Metric) 23 mm