Learn More
Memmert™ Entry Port 23 mm for IN55, Custom TBC Back Side
Product Code. 17597825 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN55 Incubator
Unit Size:
Each
Product Code. 17597825
Brand: Memmert™ IN55F6
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IN55 Incubator
Type Entry Port
Diameter (Metric) 23 mm