Learn More
Memmert™ Entry Port 23 mm for IF260, Standard Position Left Side Center/Top
Product Code. 17556815 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF260 Incubator
Unit Size:
Each
Product Code. 17556815
Brand: Memmert™ IF260F1
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Incubators.
  • Entry port with 23 mm diameter
  • Standard position left side center/top
Specifications
For Use With (Equipment) IF260 Incubator
Type Entry Port
Diameter (Metric) 23 mm